Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567843_m1
          See other TNNI1 GE Assays ›
          Gene Symbol:
          Tnni1
          Gene Name:
          troponin I type 1 (skeletal, slow)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 52624856 - 52637297 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567843_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnni1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017184.1 NP_058880.1 4 - 5 232 129
          XM_006249862.2 XP_006249924.1 3 - 4 1739 129
          XM_006249864.1 XP_006249926.1 3 - 4 318 129
          XM_006249865.2 XP_006249927.1 6 - 7 361 129
          XM_006249866.2 XP_006249928.1 6 - 7 292 129
          XM_006249868.2 XP_006249930.1 4 - 5 327 129
          XM_006249869.2 XP_006249931.1 4 - 5 251 129
          GenBank mRNA FQ214889.1 - 6 - 7 339 129
          FQ215520.1 - 6 - 7 314 129
          FQ215574.1 - 6 - 7 312 129
          FQ216094.1 - 6 - 7 312 129
          FQ216822.1 - 6 - 7 312 129
          FQ217411.1 - 6 - 7 312 129
          J04993.1 - 4 - 5 232 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000346.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnni1

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of muscle contraction
          transition between fast and slow fiber
          ventricular cardiac muscle tissue morphogenesis
          actin binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P577677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154474 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P577677 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P577677 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P577677 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P577677 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30487 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10764236 - -
          Affymetrix Rat Gene 1.0 ST Array 10764239 - -
          Affymetrix Rat Gene 1.0 ST Array 10764234 - -
          Affymetrix Rat Gene 1.0 ST Array 10764238 - -
          Affymetrix Rat Gene 1.0 ST Array 10764237 - -
          Affymetrix Rat Gene 1.0 ST Array 10764235 - -
          Affymetrix Rat Gene 1.1 ST Array 10764233 - -
          Rat Genome 230 2.0 Array 1386873_at - -
          Rat Genome 230 2.0 Array 1371339_PM_at - -
          Rat Genome 230 2.0 Array 1386873_PM_at - -
          Rat Genome 230 2.0 Array 1371339_at - -
          Affymetrix HT Rat Focus Array 1386873_PM_at - -
          Affymetrix HT Rat Focus Array 1386873_at - -
          Affymetrix Rat Clariom S Assay PSR1300002283.rn.1 TC1300000346.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300002284.rn.1 TC1300000346.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300002285.rn.1 TC1300000346.rn.1 RTA TC
          Rat Expression Set 230 1371339_PM_at - -
          Rat Expression Set 230 1386873_PM_at - -
          Rat Expression Set 230 1386873_at - -
          Rat Expression Set 230 1371339_at - -
          Affymetrix Rat Gene 1.0 ST Array 10764233 - -
          Rat Genome U34 Set J04993_at - -
          Rat Genome U34 Set rc_AA859335_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21437088 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354309 - -
          RatRef-12 Expression BeadChip ILMN_58172 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline