Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571829_m1
          See other MYL1 GE Assays ›
          Gene Symbol:
          Myl1
          Gene Name:
          myosin, light chain 1
          Gene Aliases:
          MLC3-f, MLCf, Mcl3, Mlc1, Mlc3
          Chromosome Location:
          Chr.9: 73937820 - 73958480 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571829_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077656.1 NP_001071124.1 3 - 4 404 87
          NM_020104.2 NP_064489.1 3 - 4 222 87
          XM_008767294.1 XP_008765516.1 2 - 3 352 87
          XM_008767295.1 XP_008765517.1 2 - 3 473 87
          GenBank mRNA CR755322.1 - 3 - 4 227 87
          CR756356.1 - 3 - 4 222 87
          FQ214580.1 - 3 - 4 531 87
          FQ214604.1 - 3 - 4 449 87
          FQ214608.1 - 3 - 4 409 87
          FQ214631.1 - 3 - 4 391 87
          FQ214638.1 - 3 - 4 498 87
          FQ214656.1 - 3 - 4 240 87
          FQ214675.1 - 3 - 4 498 87
          FQ214712.1 - 3 - 4 412 87
          FQ214723.1 - 3 - 4 412 87
          FQ214727.1 - 3 - 4 412 87
          FQ214742.1 - 3 - 4 412 87
          FQ214939.1 - 3 - 4 409 87
          FQ215002.1 - 3 - 4 498 87
          FQ215050.1 - 3 - 4 412 87
          FQ215083.1 - 3 - 4 409 87
          FQ215086.1 - 3 - 4 242 87
          FQ215201.1 - 3 - 4 409 87
          FQ215229.1 - 3 - 4 503 87
          FQ215325.1 - 3 - 4 498 87
          FQ215340.1 - 3 - 4 240 87
          FQ215418.1 - 3 - 4 498 87
          FQ215435.1 - 3 - 4 409 87
          FQ215492.1 - 3 - 4 391 87
          FQ215500.1 - 2 - 3 988 87
          FQ215557.1 - 3 - 4 498 87
          FQ215563.1 - 3 - 4 409 87
          FQ215578.1 - 3 - 4 498 87
          FQ215610.1 - 3 - 4 412 87
          FQ215748.1 - 2 - 3 471 87
          FQ215766.1 - 3 - 4 498 87
          FQ215774.1 - 3 - 4 409 87
          FQ215820.1 - 3 - 4 467 87
          FQ215848.1 - 3 - 4 409 87
          FQ215875.1 - 3 - 4 409 87
          FQ215911.1 - 3 - 4 503 87
          FQ215933.1 - 3 - 4 412 87
          FQ216034.1 - 3 - 4 412 87
          FQ216109.1 - 3 - 4 409 87
          FQ216124.1 - 3 - 4 503 87
          FQ216181.1 - 3 - 4 391 87
          FQ216192.1 - 3 - 4 498 87
          FQ216213.1 - 4 - 5 406 87
          FQ216218.1 - 3 - 4 409 87
          FQ216229.1 - 3 - 4 679 87
          FQ216238.1 - 3 - 4 498 87
          FQ216308.1 - 3 - 4 240 87
          FQ216407.1 - 3 - 4 580 87
          FQ216452.1 - 3 - 4 409 87
          FQ216455.1 - 3 - 4 413 87
          FQ216484.1 - 3 - 4 391 87
          FQ216496.1 - 3 - 4 412 87
          FQ216525.1 - 3 - 4 412 87
          FQ216543.1 - 3 - 4 409 87
          FQ216556.1 - 3 - 4 498 87
          FQ216567.1 - 3 - 4 409 87
          FQ216631.1 - 3 - 4 412 87
          FQ216704.1 - 3 - 4 412 87
          FQ216738.1 - 3 - 4 240 87
          FQ217074.1 - 3 - 4 239 87
          FQ217103.1 - 3 - 4 240 87
          FQ217157.1 - 2 - 3 215 87
          FQ217192.1 - 3 - 4 240 87
          FQ217218.1 - 3 - 4 240 87
          FQ217243.1 - 3 - 4 239 87
          FQ217303.1 - 3 - 4 391 87
          FQ217323.1 - 3 - 4 240 87
          FQ217387.1 - 3 - 4 240 87
          FQ217394.1 - 3 - 4 342 87
          FQ217427.1 - 3 - 4 245 87
          FQ217481.1 - 3 - 4 240 87
          FQ217753.1 - 3 - 4 245 87
          FQ217756.1 - 3 - 4 240 87
          FQ217889.1 - 3 - 4 240 87
          FQ217979.1 - 3 - 4 240 87
          FQ218029.1 - 3 - 4 240 87
          FQ218049.1 - 3 - 4 240 87
          FQ223632.1 - 3 - 4 412 87
          FQ223641.1 - 3 - 4 498 87
          FQ223687.1 - 3 - 4 240 87
          FQ223755.1 - 3 - 4 408 87
          FQ223779.1 - 3 - 4 240 87
          FQ223827.1 - 3 - 4 412 87
          FQ223880.1 - 3 - 4 310 87
          FQ223949.1 - 3 - 4 238 87
          FQ223984.1 - 3 - 4 240 87
          FQ224047.1 - 3 - 4 240 87
          FQ224050.1 - 3 - 4 240 87
          FQ224087.1 - 3 - 4 240 87
          FQ224112.1 - 2 - 3 215 87
          FQ224155.1 - 3 - 4 245 87
          FQ224171.1 - 3 - 4 409 87
          FQ224172.1 - 3 - 4 240 87
          FQ224254.1 - 3 - 4 240 87
          FQ224277.1 - 3 - 4 412 87
          FQ224321.1 - 3 - 4 244 87
          FQ224386.1 - 3 - 4 245 87
          FQ224411.1 - 3 - 4 240 87
          FQ224420.1 - 3 - 4 240 87
          FQ224452.1 - 3 - 4 315 87
          FQ224519.1 - 3 - 4 409 87
          FQ224609.1 - 4 - 5 268 87
          FQ224663.1 - 3 - 4 240 87
          FQ224726.1 - 3 - 4 342 87
          FQ224796.1 - 3 - 4 240 87
          FQ224799.1 - 3 - 4 245 87
          FQ224927.1 - 3 - 4 498 87
          FQ224931.1 - 3 - 4 409 87
          FQ224934.1 - 3 - 4 407 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002140.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myl1

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          cardiac muscle contraction
          calcium ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0900017600.rn.1 TC0900002140.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline