Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00576900_m1
          See other MCAM GE Assays ›
          Gene Symbol:
          Mcam
          Gene Name:
          melanoma cell adhesion molecule
          Gene Aliases:
          CD146, Muc18
          Chromosome Location:
          Chr.8: 48472824 - 48481005 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00576900_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mcam
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034009.1 NP_001029181.1 5 - 6 579 84
          NM_023983.3 NP_076473.2 5 - 6 579 84
          GenBank mRNA AB035506.1 - 5 - 6 577 84
          AB035507.1 - 5 - 6 577 84
          BC070916.1 - 5 - 6 579 7105099 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000555.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mcam

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          glomerular filtration
          cell adhesion
          positive regulation of cell migration
          vascular wound healing
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P317388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956316 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32057 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P317388 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P317388 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P317388 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P463504 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P317388 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700290569 - -
          Agilent Rat cDNA Microarray (G4105A) 700290569 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10909450 - -
          Affymetrix Rat Gene 1.0 ST Array 10909449 - -
          Affymetrix Rat Gene 1.0 ST Array 10909447 - -
          Affymetrix Rat Gene 1.0 ST Array 10909461 - -
          Affymetrix Rat Gene 1.0 ST Array 10909462 - -
          Affymetrix Rat Gene 1.0 ST Array 10909452 - -
          Affymetrix Rat Gene 1.0 ST Array 10909459 - -
          Affymetrix Rat Gene 1.0 ST Array 10909458 - -
          Affymetrix Rat Gene 1.0 ST Array 10909454 - -
          Affymetrix Rat Gene 1.0 ST Array 10909457 - -
          Affymetrix Rat Gene 1.0 ST Array 10909455 - -
          Affymetrix Rat Gene 1.0 ST Array 10909448 - -
          Affymetrix Rat Gene 1.0 ST Array 10909456 - -
          Affymetrix Rat Gene 1.0 ST Array 10909460 - -
          Affymetrix Rat Gene 1.0 ST Array 10909451 - -
          Affymetrix Rat Gene 1.0 ST Array 10909453 - -
          Affymetrix Rat Gene 1.1 ST Array 10909446 - -
          Rat Genome 230 2.0 Array 1369793_a_at - -
          Rat Genome 230 2.0 Array 1374933_PM_at - -
          Rat Genome 230 2.0 Array 1369793_PM_a_at - -
          Rat Genome 230 2.0 Array 1374933_at - -
          Affymetrix HT Rat Focus Array 1374933_PM_at - -
          Affymetrix HT Rat Focus Array 1374933_at - -
          Affymetrix HT Rat Focus Array 1369793_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369793_a_at - -
          Affymetrix Rat Clariom S Assay PSR0800003432.rn.1 TC0800000555.rn.1 RTA TC
          Rat Expression Set 230 1374933_PM_at - -
          Rat Expression Set 230 1374933_at - -
          Rat Expression Set 230 1369793_a_at - -
          Rat Expression Set 230 1369793_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10909446 - -
          Rat Genome U34 Set rc_AA859350_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20741650 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357064 - -
          RatRef-12 Expression BeadChip ILMN_60488 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline