Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579457_m1
          See other RBL2 GE Assays ›
          Gene Symbol:
          Rbl2
          Gene Name:
          retinoblastoma-like 2
          Gene Aliases:
          Rb2
          Chromosome Location:
          Chr.19: 17300088 - 17346863 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579457_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031094.1 NP_112356.1 5 - 6 825 85
          XM_006255199.2 XP_006255261.1 5 - 6 879 85
          XM_006255200.2 XP_006255262.1 5 - 6 880 85
          XM_006255201.2 XP_006255263.1 2 - 3 959 85
          GenBank mRNA D55627.1 - 5 - 6 825 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001076.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbl2

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          chromatin modification
          regulation of lipid kinase activity
          regulation of cell cycle
          DNA binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840041 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89295 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84921 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91973 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57911 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71463 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15508 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15508 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15508 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P264749 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15508 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700933578 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36349 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14682 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12709 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26725 - -
          Agilent Rat cDNA Microarray (G4105A) 700933578 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10809584 - -
          Affymetrix Rat Gene 1.0 ST Array 10809590 - -
          Affymetrix Rat Gene 1.0 ST Array 10809596 - -
          Affymetrix Rat Gene 1.0 ST Array 10809588 - -
          Affymetrix Rat Gene 1.0 ST Array 10809593 - -
          Affymetrix Rat Gene 1.0 ST Array 10809594 - -
          Affymetrix Rat Gene 1.0 ST Array 10809595 - -
          Affymetrix Rat Gene 1.0 ST Array 10809602 - -
          Affymetrix Rat Gene 1.0 ST Array 10809599 - -
          Affymetrix Rat Gene 1.0 ST Array 10809591 - -
          Affymetrix Rat Gene 1.0 ST Array 10809589 - -
          Affymetrix Rat Gene 1.0 ST Array 10809586 - -
          Affymetrix Rat Gene 1.0 ST Array 10809587 - -
          Affymetrix Rat Gene 1.0 ST Array 10809592 - -
          Affymetrix Rat Gene 1.0 ST Array 10809598 - -
          Affymetrix Rat Gene 1.0 ST Array 10809582 - -
          Affymetrix Rat Gene 1.0 ST Array 10809597 - -
          Affymetrix Rat Gene 1.0 ST Array 10809585 - -
          Affymetrix Rat Gene 1.0 ST Array 10809603 - -
          Affymetrix Rat Gene 1.0 ST Array 10809583 - -
          Affymetrix Rat Gene 1.0 ST Array 10809601 - -
          Affymetrix Rat Gene 1.0 ST Array 10809600 - -
          Affymetrix Rat Gene 1.1 ST Array 10809581 - -
          Rat Genome 230 2.0 Array 1368403_at - -
          Rat Genome 230 2.0 Array 1368403_PM_at - -
          Affymetrix HT Rat Focus Array 1368403_at - -
          Affymetrix HT Rat Focus Array 1368403_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900008618.rn.1 TC1900001076.rn.1 RTA TC
          Rat Expression Set 230 1368403_at - -
          Rat Expression Set 230 1368403_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10809581 - -
          Rat Genome U34 Set rc_AI227715_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20712530 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360394 - -
          RatRef-12 Expression BeadChip ILMN_59080 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline