Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585595_m1
          See other PICALM GE Assays ›
          Gene Symbol:
          Picalm
          Gene Name:
          phosphatidylinositol binding clathrin assembly protein
          Gene Aliases:
          Calm
          Chromosome Location:
          Chr.1: 154377234 - 154458922 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585595_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Picalm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053554.2 NP_446006.1 19 - 20 1928 91
          XM_006229676.2 XP_006229738.1 20 - 21 2257 91
          XM_006229677.2 XP_006229739.1 19 - 20 2233 91
          XM_006229678.2 XP_006229740.1 19 - 20 2218 91
          XM_006229683.2 XP_006229745.1 19 - 20 2107 91
          XM_006229684.2 XP_006229746.1 19 - 20 2092 91
          XM_006229685.2 XP_006229747.1 18 - 19 2068 91
          GenBank mRNA AF041373.1 - 18 - 19 1800 91
          AF041374.1 - 19 - 20 1929 91
          AY724477.1 - 18 - 19 1793 91
          BC100142.1 - 19 - 20 1989 7456159 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001998.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Picalm

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          receptor-mediated endocytosis
          axonogenesis
          cell proliferation
          negative regulation of gene expression
          synaptic vesicle maturation
          vesicle-mediated transport
          endosomal transport
          hemopoiesis
          regulation of endocytosis
          receptor internalization
          regulation of protein localization
          cargo loading into vesicle
          positive regulation of transcription, DNA-templated
          negative regulation of receptor-mediated endocytosis
          clathrin coat assembly
          dendrite morphogenesis
          regulation of protein transport
          iron ion homeostasis
          clathrin-mediated endocytosis
          iron ion import into cell
          positive regulation of neuron death
          positive regulation of beta-amyloid formation
          regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          positive regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          negative regulation of metalloendopeptidase activity involved in amyloid precursor protein catabolic process
          protein binding
          1-phosphatidylinositol binding
          SH3 domain binding
          clathrin binding
          clathrin heavy chain binding
          clathrin adaptor activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P001525 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503709 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50989 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112755 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163046 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162339 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84508 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137370 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72989 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79027 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6703 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23318 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001525 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12852 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12852 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001525 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12852 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P001525 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P360795 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700601617 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32609 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2740 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42231 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56929 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40900 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46512 - -
          Agilent Rat cDNA Microarray (G4105A) 700601617 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10708584 - -
          Affymetrix Rat Gene 1.0 ST Array 10708573 - -
          Affymetrix Rat Gene 1.0 ST Array 10708583 - -
          Affymetrix Rat Gene 1.0 ST Array 10708579 - -
          Affymetrix Rat Gene 1.0 ST Array 10708580 - -
          Affymetrix Rat Gene 1.0 ST Array 10708566 - -
          Affymetrix Rat Gene 1.0 ST Array 10708574 - -
          Affymetrix Rat Gene 1.0 ST Array 10708581 - -
          Affymetrix Rat Gene 1.0 ST Array 10708582 - -
          Affymetrix Rat Gene 1.0 ST Array 10708570 - -
          Affymetrix Rat Gene 1.0 ST Array 10708576 - -
          Affymetrix Rat Gene 1.0 ST Array 10708586 - -
          Affymetrix Rat Gene 1.0 ST Array 10708568 - -
          Affymetrix Rat Gene 1.0 ST Array 10708572 - -
          Affymetrix Rat Gene 1.0 ST Array 10708578 - -
          Affymetrix Rat Gene 1.0 ST Array 10708577 - -
          Affymetrix Rat Gene 1.0 ST Array 10708567 - -
          Affymetrix Rat Gene 1.0 ST Array 10708569 - -
          Affymetrix Rat Gene 1.0 ST Array 10708571 - -
          Affymetrix Rat Gene 1.0 ST Array 10708575 - -
          Affymetrix Rat Gene 1.0 ST Array 10708585 - -
          Affymetrix Rat Gene 1.1 ST Array 10708565 - -
          Rat Genome 230 2.0 Array 1369452_PM_a_at - -
          Rat Genome 230 2.0 Array 1373877_at - -
          Rat Genome 230 2.0 Array 1369452_a_at - -
          Rat Genome 230 2.0 Array 1373877_PM_at - -
          Affymetrix HT Rat Focus Array 1369452_a_at - -
          Affymetrix HT Rat Focus Array 1369452_PM_a_at - -
          Affymetrix HT Rat Focus Array 1373877_PM_at - -
          Affymetrix HT Rat Focus Array 1373877_at - -
          Affymetrix Rat Clariom S Assay PSR0100015530.rn.1 TC0100001998.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100015532.rn.1 TC0100001998.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100015533.rn.1 TC0100001998.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100015534.rn.1 TC0100001998.rn.1 RTA TC
          Rat Expression Set 230 1369452_PM_a_at - -
          Rat Expression Set 230 1369452_a_at - -
          Rat Expression Set 230 1373877_PM_at - -
          Rat Expression Set 230 1373877_at - -
          Affymetrix Rat Gene 1.0 ST Array 10708565 - -
          Rat Genome U34 Set AF041373_s_at - -
          Rat Genome U34 Set rc_AI169149_at - -
          Rat Genome U34 Set AF041373_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22186588 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374519 - -
          RatRef-12 Expression BeadChip ILMN_56526 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline