Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00673281_m1
          See other BTC GE Assays ›
          Gene Symbol:
          Btc
          Gene Name:
          betacellulin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 18231854 - 18270621 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00673281_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Btc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022256.2 NP_071592.1 324 69
          GenBank mRNA AB028862.1 - 324 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000268.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Btc

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          epidermal growth factor receptor signaling pathway
          positive regulation of cell proliferation
          cell differentiation
          positive regulation of urine volume
          negative regulation of apoptotic process
          positive regulation of cell differentiation
          positive regulation of mitotic nuclear division
          positive regulation of fibroblast proliferation
          epidermal growth factor receptor binding
          growth factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P043767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232632 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144399 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147994 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043767 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043767 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P043767 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12186 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8376 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55139 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10771761 - -
          Affymetrix Rat Gene 1.0 ST Array 10771764 - -
          Affymetrix Rat Gene 1.0 ST Array 10771763 - -
          Affymetrix Rat Gene 1.0 ST Array 10771762 - -
          Affymetrix Rat Gene 1.1 ST Array 10771760 - -
          Rat Genome 230 2.0 Array 1387644_at - -
          Rat Genome 230 2.0 Array 1387644_PM_at - -
          Affymetrix HT Rat Focus Array 1387644_at - -
          Affymetrix HT Rat Focus Array 1387644_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1400002148.rn.1 TC1400000268.rn.1 RTA TC
          Rat Expression Set 230 1387644_at - -
          Rat Expression Set 230 1387644_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10771760 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21495686 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372200 - -
          RatRef-12 Expression BeadChip ILMN_57060 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline