Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00695344_g1
          See other SPATA7 GE Assays ›
          Gene Symbol:
          Spata7
          Gene Name:
          spermatogenesis associated 7
          Gene Aliases:
          RSD-3, Wmp1
          Chromosome Location:
          Chr.6: 122603252 - 122648718 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00695344_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spata7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138862.3 NP_620217.2 11 - 12 1250 85
          XM_006240413.2 XP_006240475.1 10 - 11 1233 85
          XM_006240414.2 XP_006240476.1 11 - 12 1325 85
          XM_006240415.2 XP_006240477.1 9 - 10 1158 85
          XM_008764737.1 XP_008762959.1 10 - 11 1204 85
          GenBank mRNA BC078728.1 - 11 - 12 976 7114267 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001320.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spata7

          Gene Ontology Categories:

          Function(s)

          spermatogenesis
          photoreceptor cell maintenance
          protein localization to photoreceptor outer segment
          protein localization to photoreceptor connecting cilium

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P102661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954227 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102079 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127852 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44810 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134568 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P102661 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P102661 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P102661 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P102661 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80585 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61461 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65751 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61244 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46061 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55818 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10886281 - -
          Affymetrix Rat Gene 1.0 ST Array 10886293 - -
          Affymetrix Rat Gene 1.0 ST Array 10886294 - -
          Affymetrix Rat Gene 1.0 ST Array 10886282 - -
          Affymetrix Rat Gene 1.0 ST Array 10886284 - -
          Affymetrix Rat Gene 1.0 ST Array 10886286 - -
          Affymetrix Rat Gene 1.0 ST Array 10886291 - -
          Affymetrix Rat Gene 1.0 ST Array 10886288 - -
          Affymetrix Rat Gene 1.0 ST Array 10886292 - -
          Affymetrix Rat Gene 1.0 ST Array 10886285 - -
          Affymetrix Rat Gene 1.0 ST Array 10886283 - -
          Affymetrix Rat Gene 1.0 ST Array 10886290 - -
          Affymetrix Rat Gene 1.0 ST Array 10886289 - -
          Affymetrix Rat Gene 1.0 ST Array 10886287 - -
          Affymetrix Rat Gene 1.1 ST Array 10886280 - -
          Rat Genome 230 2.0 Array 1370398_at - -
          Rat Genome 230 2.0 Array 1370398_PM_at - -
          Affymetrix HT Rat Focus Array 1370398_at - -
          Affymetrix HT Rat Focus Array 1370398_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600009105.rn.1 TC0600001320.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009108.rn.1 TC0600001320.rn.1 RTA TC
          Rat Expression Set 230 1370398_PM_at - -
          Rat Expression Set 230 1370398_at - -
          Affymetrix Rat Gene 1.0 ST Array 10886280 - -
          Rat Genome U34 Set rc_AI045519_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21641591 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376540 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline