Search
Search
| Gene Symbol: Ldhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012595.1 | NP_036727.1 | 7 - 8 | 863 | 69 | |
| XM_006237615.2 | XP_006237677.1 | 8 - 9 | 1034 | 69 | ||
| GenBank mRNA | BC059149.1 | - | 7 - 8 | 920 | 6919339 | 69 |
| FQ214847.1 | - | 7 - 8 | 910 | 69 | ||
| FQ215687.1 | - | 7 - 8 | 926 | 69 | ||
| U07181.1 | - | 7 - 8 | 863 | 69 | ||
Set Membership: |
|