Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821402_g1
          See other DBI GE Assays ›
          Gene Symbol:
          Dbi
          Gene Name:
          diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein)
          Gene Aliases:
          Acbp, Acoabp3, Ep, Odn, RNACOABP3, Ttn
          Chromosome Location:
          Chr.13: 36147667 - 36156076 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821402_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dbi
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031853.4 NP_114054.1 2 - 3 244 58
          GenBank mRNA AY383701.1 - 424 58
          BC084717.1 - 1 - 2 131 7310272 58
          FQ209769.1 - 2 - 3 170 58
          FQ209835.1 - 2 - 3 192 58
          FQ209938.1 - 2 - 3 187 58
          FQ210140.1 - 2 - 3 192 58
          FQ210165.1 - 2 - 3 187 58
          FQ210336.1 - 2 - 3 189 58
          FQ210781.1 - 2 - 3 219 58
          FQ210997.1 - 2 - 3 189 58
          FQ211081.1 - 2 - 3 192 58
          FQ211136.1 - 2 - 3 192 58
          FQ211192.1 - 2 - 3 192 58
          FQ211339.1 - 2 - 3 193 58
          FQ211377.1 - 1 - 2 113 58
          FQ213527.1 - 2 - 3 193 58
          FQ214572.1 - 2 - 3 192 58
          FQ217714.1 - 2 - 3 193 58
          FQ218164.1 - 2 - 3 211 58
          FQ218680.1 - 2 - 3 192 58
          FQ219739.1 - 2 - 3 192 58
          FQ221153.1 - 2 - 3 187 58
          FQ221871.1 - 2 - 3 193 58
          FQ222052.1 - 2 - 3 192 58
          FQ222378.1 - 2 - 3 194 58
          FQ222416.1 - 2 - 3 187 58
          FQ222549.1 - 2 - 3 179 58
          FQ222564.1 - 2 - 3 193 58
          FQ222579.1 - 2 - 3 223 58
          FQ223167.1 - 2 - 3 187 58
          FQ223413.1 - 2 - 3 192 58
          FQ223492.1 - 2 - 3 193 58
          FQ223602.1 - 2 - 3 150 58
          FQ224361.1 - 2 - 3 192 58
          FQ224797.1 - 2 - 3 193 58
          FQ229114.1 - 2 - 3 187 58
          M14201.1 - 2 - 3 244 58
          M20268.1 - 2 - 3 133 58

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dbi

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          hair follicle development
          acyl-CoA metabolic process
          triglyceride metabolic process
          steroid biosynthetic process
          brain development
          learning or memory
          glial cell proliferation
          protein palmitoylation
          lateral ventricle development
          pancreatic juice secretion
          cellular response to nutrient
          negative regulation of fatty acid beta-oxidation
          regulation of synaptic transmission, GABAergic
          phosphatidylcholine acyl-chain remodeling
          skin development
          positive regulation of lipid biosynthetic process
          positive regulation of release of sequestered calcium ion into cytosol
          long-term synaptic potentiation
          fatty-acyl-CoA binding
          protein binding
          lipid binding
          benzodiazepine receptor binding
          long-chain fatty acyl-CoA binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14621 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14621 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14621 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14621 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P141377 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14621 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700487123 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39585 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58919 - -
          Agilent Rat cDNA Microarray (G4105A) 700487123 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10931155 - -
          Affymetrix Rat Gene 1.0 ST Array 10931158 - -
          Affymetrix Rat Gene 1.0 ST Array 10931157 - -
          Affymetrix Rat Gene 1.1 ST Array 10931154 - -
          Rat Genome 230 2.0 Array 1370235_PM_at - -
          Rat Genome 230 2.0 Array 1370235_at - -
          Affymetrix HT Rat Focus Array 1370235_at - -
          Affymetrix HT Rat Focus Array 1370235_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300008441.rn.1 TC1300001270.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300008443.rn.1 TC1300001270.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300008440.rn.1 TC1300001270.rn.1 RTA TC
          Rat Expression Set 230 1370235_at - -
          Rat Expression Set 230 1370235_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10931154 - -
          Rat Genome U34 Set rc_AI010581_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22265198 - -
          ABI Rat Genome Survey Microarray 22425679 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354106 - -
          RatRef-12 Expression BeadChip ILMN_60315 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline