Search
Search
| Gene Symbol: Foxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031633.3 | NP_113821.2 | 6 - 7 | 1098 | 69 | |
| XM_006237416.2 | XP_006237478.1 | 4 - 5 | 1038 | 69 | ||
| XM_006237417.2 | XP_006237479.1 | 4 - 5 | 1038 | 69 | ||
| XM_006237418.2 | XP_006237480.1 | 5 - 6 | 1062 | 69 | ||
| XM_006237421.2 | XP_006237483.1 | 5 - 6 | 1062 | 69 | ||
| GenBank mRNA | U83112.1 | - | 6 - 7 | 1098 | 69 | |
Set Membership: |
|