Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01401365_m1
          See other ZFP521 GE Assays ›
          Gene Symbol:
          Zfp521
          Gene Name:
          zinc finger protein 521
          Gene Aliases:
          Znf521
          Chromosome Location:
          Chr.18: 5022042 - 5314639 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01401365_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp521
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107403.1 NP_001100873.1 6 - 7 3913 73
          XM_008771948.1 XP_008770170.1 6 - 7 4019 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001052.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Process(es)

          nucleic acid binding
          protein domain specific binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P090846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122579 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158912 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165273 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6180 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161185 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54489 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100170 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160200 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63549 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115513 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61157 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174241 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176200 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21724 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86691 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56711 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176983 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P090846 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P090846 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P090846 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P515809 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35713 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100005 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102618 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85047 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99823 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10803280 - -
          Affymetrix Rat Gene 1.0 ST Array 10803277 - -
          Affymetrix Rat Gene 1.0 ST Array 10803279 - -
          Affymetrix Rat Gene 1.0 ST Array 10803278 - -
          Affymetrix Rat Gene 1.1 ST Array 10803276 - -
          Rat Genome 230 2.0 Array 1393944_PM_at - -
          Rat Genome 230 2.0 Array 1393944_at - -
          Affymetrix Rat Clariom S Assay PSR1800007455.rn.1 TC1800001052.rn.1 RTA TC
          Rat Expression Set 230 1393944_PM_at - -
          Rat Expression Set 230 1393944_at - -
          Affymetrix Rat Gene 1.0 ST Array 10803276 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20718173 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353751 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline