Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01405402_m1
See other TBX1 GE Assays ›
Gene Symbol:
Tbx1
Gene Name:
T-box 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 86552022 - 86561647 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01405402_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Tbx1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001108322.1
NP_001101792.1
3 - 4
511
81
XM_006248619.2
XP_006248681.1
2 - 3
1293
81
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1100000986.rn.1
Set Membership:
Amplicon length between 71 and 85
Rattus norvegicus
Probe spans exons
Panther Classification:
Molecular Function -
Rel homology transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
blood vessel development
cell fate specification
neural crest cell migration
positive regulation of protein phosphorylation
lymph vessel development
blood vessel remodeling
positive regulation of mesenchymal cell proliferation
heart morphogenesis
outflow tract septum morphogenesis
outflow tract morphogenesis
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
determination of left/right symmetry
pattern specification process
mesoderm development
heart development
muscle organ development
sensory perception of sound
biological_process
cell proliferation
positive regulation of cell proliferation
anterior/posterior pattern specification
vagus nerve morphogenesis
epithelial cell differentiation
thyroid gland development
social behavior
aorta morphogenesis
ear morphogenesis
inner ear morphogenesis
outer ear morphogenesis
middle ear morphogenesis
odontogenesis of dentin-containing tooth
muscle cell fate commitment
positive regulation of MAPK cascade
tongue morphogenesis
cellular response to fibroblast growth factor stimulus
negative regulation of cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
blood vessel morphogenesis
thymus development
muscle organ morphogenesis
embryonic cranial skeleton morphogenesis
embryonic viscerocranium morphogenesis
semicircular canal morphogenesis
artery morphogenesis
positive regulation of epithelial cell proliferation
parathyroid gland development
soft palate development
pharyngeal system development
face morphogenesis
muscle tissue morphogenesis
coronary artery morphogenesis
enamel mineralization
cellular response to retinoic acid
otic vesicle morphogenesis
positive regulation of secondary heart field cardioblast proliferation
cochlea morphogenesis
mesenchymal cell apoptotic process
regulation of organ morphogenesis
positive regulation of tongue muscle cell differentiation
negative regulation of mesenchymal cell apoptotic process
RNA polymerase II intronic transcription regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
protein homodimerization activity
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR1100005612.rn.1
TC1100000986.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1100005614.rn.1
TC1100000986.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline