Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01424337_m1
          See other PDK3 GE Assays ›
          Gene Symbol:
          Pdk3
          Gene Name:
          pyruvate dehydrogenase kinase, isozyme 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 62836131 - 62904114 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01424337_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdk3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106581.1 NP_001100051.1 11 - 12 1258 61
          XM_006257018.2 XP_006257080.1 10 - 11 1325 61
          XM_006257019.2 XP_006257081.1 10 - 11 1336 61
          XM_006257020.2 XP_006257082.1 10 - 11 1327 61
          GenBank mRNA BC169078.1 - 10 - 11 1272 5598223 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001861.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          regulation of acetyl-CoA biosynthetic process from pyruvate
          regulation of glucose metabolic process
          peptidyl-serine phosphorylation
          peroxisome proliferator activated receptor signaling pathway
          cellular response to glucose stimulus
          cellular response to fatty acid
          hypoxia-inducible factor-1alpha signaling pathway
          regulation of reactive oxygen species metabolic process
          molecular_function
          protein kinase activity
          protein serine/threonine kinase activity
          pyruvate dehydrogenase (acetyl-transferring) kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P991682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144536 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132206 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135469 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114261 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101658 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173126 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149745 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53244 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P991682 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P991682 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P991682 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P443343 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P991682 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32064 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51818 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10938389 - -
          Affymetrix Rat Gene 1.0 ST Array 10938386 - -
          Affymetrix Rat Gene 1.0 ST Array 10938394 - -
          Affymetrix Rat Gene 1.0 ST Array 10938392 - -
          Affymetrix Rat Gene 1.0 ST Array 10938393 - -
          Affymetrix Rat Gene 1.0 ST Array 10938385 - -
          Affymetrix Rat Gene 1.0 ST Array 10938388 - -
          Affymetrix Rat Gene 1.0 ST Array 10938391 - -
          Affymetrix Rat Gene 1.0 ST Array 10938390 - -
          Affymetrix Rat Gene 1.0 ST Array 10938387 - -
          Affymetrix Rat Gene 1.1 ST Array 10938384 - -
          Rat Genome 230 2.0 Array 1372229_PM_at - -
          Rat Genome 230 2.0 Array 1384315_PM_at - -
          Rat Genome 230 2.0 Array 1372229_at - -
          Rat Genome 230 2.0 Array 1384315_at - -
          Affymetrix HT Rat Focus Array 1372229_at - -
          Affymetrix HT Rat Focus Array 1372229_PM_at - -
          Affymetrix HT Rat Focus Array 1384315_PM_at - -
          Affymetrix HT Rat Focus Array 1384315_at - -
          Affymetrix Rat Clariom S Assay PSR0X00011737.rn.1 TC0X00001861.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00011738.rn.1 TC0X00001861.rn.1 RTA TC
          Rat Expression Set 230 1372229_PM_at - -
          Rat Expression Set 230 1372229_at - -
          Rat Expression Set 230 1384315_at - -
          Rat Expression Set 230 1384315_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10938384 - -
          Rat Genome U34 Set rc_AI231465_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20714771 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365682 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline