Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01429720_g1
          See other IGHA GE Assays ›
          Gene Symbol:
          Igha
          Gene Name:
          immunoglobulin heavy chain, alpha
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 138054656 - 138056613 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01429720_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igha
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AJ510151.1 - 1 - 2 308 60
          BC087057.1 - 752 60
          BC087137.1 - 797 60
          BC088269.1 - 726 60
          BC089840.1 - 792 60
          BC089851.1 - 728 60
          BC091243.1 - 821 60
          BC091247.1 - 794 60
          BC092592.1 - 762 60
          BC160870.1 - 759 60

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          immunoglobulin

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P147572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869712 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96323 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89357 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35311 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72931 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61785 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23881 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37253 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165557 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94582 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83450 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67296 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106711 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107036 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158449 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16179 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139816 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26548 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P147572 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P147572 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P147572 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66492 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54091 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9395 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96335 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5360 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56509 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50100 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67951 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49488 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81265 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15688 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82609 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17957 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34397 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83636 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10887664 - -
          Affymetrix Rat Gene 1.0 ST Array 10892663 - -
          Affymetrix Rat Gene 1.0 ST Array 10887659 - -
          Affymetrix Rat Gene 1.0 ST Array 10892514 - -
          Affymetrix Rat Gene 1.0 ST Array 10887658 - -
          Affymetrix Rat Gene 1.0 ST Array 10892664 - -
          Affymetrix Rat Gene 1.0 ST Array 10892412 - -
          Affymetrix Rat Gene 1.0 ST Array 10892642 - -
          Affymetrix Rat Gene 1.0 ST Array 10892512 - -
          Affymetrix Rat Gene 1.0 ST Array 10892413 - -
          Affymetrix Rat Gene 1.0 ST Array 10892491 - -
          Affymetrix Rat Gene 1.0 ST Array 10892504 - -
          Affymetrix Rat Gene 1.0 ST Array 10887642 - -
          Affymetrix Rat Gene 1.0 ST Array 10892441 - -
          Affymetrix Rat Gene 1.0 ST Array 10892574 - -
          Affymetrix Rat Gene 1.0 ST Array 10892442 - -
          Affymetrix Rat Gene 1.0 ST Array 10892523 - -
          Affymetrix Rat Gene 1.0 ST Array 10892411 - -
          Affymetrix Rat Gene 1.0 ST Array 10887648 - -
          Affymetrix Rat Gene 1.0 ST Array 10892576 - -
          Affymetrix Rat Gene 1.0 ST Array 10887654 - -
          Affymetrix Rat Gene 1.0 ST Array 10892641 - -
          Affymetrix Rat Gene 1.0 ST Array 10887649 - -
          Affymetrix Rat Gene 1.0 ST Array 10892410 - -
          Affymetrix Rat Gene 1.0 ST Array 10892513 - -
          Affymetrix Rat Gene 1.0 ST Array 10892573 - -
          Affymetrix Rat Gene 1.0 ST Array 10892505 - -
          Affymetrix Rat Gene 1.0 ST Array 10887663 - -
          Affymetrix Rat Gene 1.1 ST Array 10892521 - -
          Affymetrix Rat Gene 1.1 ST Array 10892575 - -
          Affymetrix Rat Gene 1.1 ST Array 10892662 - -
          Affymetrix Rat Gene 1.1 ST Array 10892572 - -
          Affymetrix Rat Gene 1.1 ST Array 10887657 - -
          Affymetrix Rat Gene 1.1 ST Array 10892409 - -
          Affymetrix Rat Gene 1.1 ST Array 10892509 - -
          Affymetrix Rat Gene 1.1 ST Array 10892640 - -
          Rat Genome 230 2.0 Array 1371284_PM_at - -
          Rat Genome 230 2.0 Array 1371284_at - -
          Rat Genome 230 2.0 Array 1374334_PM_at - -
          Rat Genome 230 2.0 Array 1371262_at - -
          Rat Genome 230 2.0 Array 1374334_at - -
          Rat Genome 230 2.0 Array 1371262_PM_at - -
          Affymetrix HT Rat Focus Array 1371284_at - -
          Affymetrix HT Rat Focus Array 1371284_PM_at - -
          Rat Expression Set 230 1371262_PM_at - -
          Rat Expression Set 230 1371284_at - -
          Rat Expression Set 230 1374334_at - -
          Rat Expression Set 230 1374334_PM_at - -
          Rat Expression Set 230 1371284_PM_at - -
          Rat Expression Set 230 1371262_at - -
          Affymetrix Rat Gene 1.0 ST Array 10892409 - -
          Affymetrix Rat Gene 1.0 ST Array 10892572 - -
          Affymetrix Rat Gene 1.0 ST Array 10892662 - -
          Affymetrix Rat Gene 1.0 ST Array 10892575 - -
          Affymetrix Rat Gene 1.0 ST Array 10892640 - -
          Rat Genome U34 Set rc_AI234828_g_at - -
          Rat Genome U34 Set rc_AI234828_at - -
          Rat Genome U34 Set L07401_f_at - -
          Rat Genome U34 Set rc_AA893565_at - -
          Rat Genome U34 Set X60290cds_s_at - -
          Rat Genome U34 Set L07402_f_at - -
          Rat Genome U34 Set rc_AA955154_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline