Search
Search
| Gene Symbol: Micu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199412.1 | NP_955444.1 | 7 - 8 | 773 | 158 | |
| XM_006256420.2 | XP_006256482.1 | 7 - 8 | 888 | 158 | ||
| XM_006256421.2 | XP_006256483.1 | 8 - 9 | 881 | 158 | ||
| XM_006256422.2 | XP_006256484.1 | 8 - 9 | 875 | 158 | ||
| GenBank mRNA | BC062075.1 | - | 7 - 8 | 773 | 5599430 | 158 |
Set Membership: |
|