Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01435145_m1
          See other IRF4 GE Assays ›
          Gene Symbol:
          Irf4
          Gene Name:
          interferon regulatory factor 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 34886746 - 34905192 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01435145_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Irf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106108.1 NP_001099578.1 8 - 9 1276 100
          XM_006253899.2 XP_006253961.1 8 - 9 1348 100
          XM_006253900.2 XP_006253962.1 7 - 8 1243 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001695.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          protein methylation
          biological_process
          peptidyl-lysine methylation
          negative regulation of toll-like receptor signaling pathway
          defense response to protozoan
          myeloid dendritic cell differentiation
          positive regulation of DNA binding
          histone H3 acetylation
          histone H4 acetylation
          positive regulation of interleukin-10 biosynthetic process
          positive regulation of interleukin-2 biosynthetic process
          positive regulation of interleukin-13 biosynthetic process
          positive regulation of interleukin-4 biosynthetic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          T-helper 17 cell lineage commitment
          regulatory region DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          molecular_function
          transcription factor activity, sequence-specific DNA binding
          protein-lysine N-methyltransferase activity
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P342426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906086 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113993 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19925 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P342426 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P342426 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P342426 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P342426 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10798176 - -
          Affymetrix Rat Gene 1.0 ST Array 10798182 - -
          Affymetrix Rat Gene 1.0 ST Array 10798178 - -
          Affymetrix Rat Gene 1.0 ST Array 10798183 - -
          Affymetrix Rat Gene 1.0 ST Array 10798181 - -
          Affymetrix Rat Gene 1.0 ST Array 10798180 - -
          Affymetrix Rat Gene 1.0 ST Array 10798179 - -
          Affymetrix Rat Gene 1.0 ST Array 10798177 - -
          Affymetrix Rat Gene 1.1 ST Array 10798175 - -
          Affymetrix Rat Clariom S Assay PSR1700011660.rn.1 TC1700001695.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700011659.rn.1 TC1700001695.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10798175 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22346257 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354706 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline