Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01441727_m1
          See other SNCB GE Assays ›
          Gene Symbol:
          Sncb
          Gene Name:
          synuclein, beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 10384472 - 10392776 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01441727_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sncb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080777.2 NP_542955.2 2 - 3 319 67
          XM_006253594.2 XP_006253656.1 3 - 4 520 67
          GenBank mRNA BC087579.1 - 2 - 3 319 7314241 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000182.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sncb

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          dopamine metabolic process
          negative regulation of neuron apoptotic process
          synapse organization
          calcium ion binding
          alpha-tubulin binding
          beta-tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P030789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689257 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83144 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030789 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030789 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P030789 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P254957 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87845 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92962 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10794141 - -
          Affymetrix Rat Gene 1.0 ST Array 10794138 - -
          Affymetrix Rat Gene 1.0 ST Array 10794137 - -
          Affymetrix Rat Gene 1.0 ST Array 10794140 - -
          Affymetrix Rat Gene 1.0 ST Array 10794139 - -
          Affymetrix Rat Gene 1.0 ST Array 10794142 - -
          Affymetrix Rat Gene 1.1 ST Array 10794136 - -
          Rat Genome 230 2.0 Array 1371568_at - -
          Rat Genome 230 2.0 Array 1369924_PM_at - -
          Rat Genome 230 2.0 Array 1369924_at - -
          Rat Genome 230 2.0 Array 1371568_PM_at - -
          Affymetrix HT Rat Focus Array 1371568_at - -
          Affymetrix HT Rat Focus Array 1369924_at - -
          Affymetrix HT Rat Focus Array 1369924_PM_at - -
          Affymetrix HT Rat Focus Array 1371568_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700001503.rn.1 TC1700000182.rn.1 RTA TC
          Rat Expression Set 230 1369924_at - -
          Rat Expression Set 230 1371568_PM_at - -
          Rat Expression Set 230 1369924_PM_at - -
          Rat Expression Set 230 1371568_at - -
          Affymetrix Rat Gene 1.0 ST Array 10794136 - -
          Rat Genome U34 Set D17764_at - -
          Rat Genome U34 Set rc_AI145381_at - -
          Rat Genome U34 Set D17764_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21846813 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354192 - -
          RatRef-12 Expression BeadChip ILMN_65288 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline