Search
Search
| Gene Symbol: Camk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021739.2 | NP_068507.2 | 15 - 16 | 1211 | 133 | |
| XM_006251170.2 | XP_006251232.1 | 15 - 16 | 1310 | 133 | ||
| XM_006251172.2 | XP_006251234.1 | 14 - 15 | 1238 | 133 | ||
| XM_006251177.2 | XP_006251239.1 | 14 - 15 | 1218 | 133 | ||
| GenBank mRNA | M16112.1 | - | 15 - 16 | 1191 | 133 | |
Set Membership: |
|