Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01447974_m1
          See other SESN3 GE Assays ›
          Gene Symbol:
          Sesn3
          Gene Name:
          sestrin 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 12823155 - 12872705 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01447974_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sesn3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108125.1 NP_001101595.1 8 - 9 1392 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000103.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s)

          response to insulin
          glucose homeostasis
          regulation of protein kinase B signaling
          regulation of response to reactive oxygen species

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P527795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869601 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14776 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80944 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107733 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96548 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63213 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76346 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144084 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P527795 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P527795 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P527795 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P527795 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P437327 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2334 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67246 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10908044 - -
          Affymetrix Rat Gene 1.0 ST Array 10908038 - -
          Affymetrix Rat Gene 1.0 ST Array 10908046 - -
          Affymetrix Rat Gene 1.0 ST Array 10908047 - -
          Affymetrix Rat Gene 1.0 ST Array 10908041 - -
          Affymetrix Rat Gene 1.0 ST Array 10908040 - -
          Affymetrix Rat Gene 1.0 ST Array 10908039 - -
          Affymetrix Rat Gene 1.0 ST Array 10908042 - -
          Affymetrix Rat Gene 1.0 ST Array 10908043 - -
          Affymetrix Rat Gene 1.0 ST Array 10908045 - -
          Affymetrix Rat Gene 1.1 ST Array 10908037 - -
          Rat Genome 230 2.0 Array 1390910_PM_at - -
          Rat Genome 230 2.0 Array 1393620_PM_at - -
          Rat Genome 230 2.0 Array 1393620_at - -
          Rat Genome 230 2.0 Array 1390910_at - -
          Affymetrix HT Rat Focus Array 1390910_PM_at - -
          Affymetrix HT Rat Focus Array 1390910_at - -
          Affymetrix HT Rat Focus Array 1393620_at - -
          Affymetrix HT Rat Focus Array 1393620_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800000591.rn.1 TC0800000103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800000592.rn.1 TC0800000103.rn.1 RTA TC
          Rat Expression Set 230 1390910_at - -
          Rat Expression Set 230 1390910_PM_at - -
          Rat Expression Set 230 1393620_PM_at - -
          Rat Expression Set 230 1393620_at - -
          Affymetrix Rat Gene 1.0 ST Array 10908037 - -
          Rat Genome U34 Set rc_AI230386_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21750480 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353619 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline