Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01448675_g1
          See other B4GALT7 GE Assays ›
          Gene Symbol:
          B4galt7
          Gene Name:
          xylosylprotein beta 1,4-galactosyltransferase, polypeptide 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 9550020 - 9558662 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01448675_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B4galt7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001031661.1 NP_001026831.1 5 - 6 874 83
          XM_008771500.1 XP_008769722.1 5 - 6 2631 83
          XM_008771501.1 XP_008769723.1 6 - 7 1074 83
          GenBank mRNA BC099103.1 - 5 - 6 874 7460148 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001315.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene B4galt7

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycosaminoglycan biosynthetic process
          proteoglycan metabolic process
          protein N-linked glycosylation
          biological_process
          extracellular fibril organization
          negative regulation of fibroblast proliferation
          molecular_function
          beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase activity
          galactosyltransferase activity
          manganese ion binding
          xylosylprotein 4-beta-galactosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P18485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10676 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18485 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18485 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P18485 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18485 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P363522 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2589 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797221 - -
          Affymetrix Rat Gene 1.0 ST Array 10797223 - -
          Affymetrix Rat Gene 1.0 ST Array 10797224 - -
          Affymetrix Rat Gene 1.0 ST Array 10797222 - -
          Affymetrix Rat Gene 1.0 ST Array 10797225 - -
          Affymetrix Rat Gene 1.0 ST Array 10797219 - -
          Affymetrix Rat Gene 1.0 ST Array 10797220 - -
          Affymetrix Rat Gene 1.1 ST Array 10797218 - -
          Rat Genome 230 2.0 Array 1379289_PM_at - -
          Rat Genome 230 2.0 Array 1379289_at - -
          Affymetrix HT Rat Focus Array 1379289_PM_at - -
          Affymetrix HT Rat Focus Array 1379289_at - -
          Affymetrix Rat Clariom S Assay PSR1700009046.rn.1 TC1700001315.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700009047.rn.1 TC1700001315.rn.1 RTA TC
          Rat Expression Set 230 1379289_PM_at - -
          Rat Expression Set 230 1379289_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797218 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352727 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline