Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01449955_m1
          See other BMP2K GE Assays ›
          Gene Symbol:
          Bmp2k
          Gene Name:
          BMP-2 inducible kinase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 14126086 - 14226823 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01449955_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bmp2k
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008770127.1 XP_008768349.1 7 - 8 1368 151

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Protein Kinase 3' Most Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          regulation of bone mineralization
          regulation of catalytic activity
          protein kinase activity
          phosphatase regulator activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P138543 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P238722 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138543 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P238722 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P238722 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138543 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P238722 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138543 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P475149 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P238722 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101491 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68722 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10775613 - -
          Affymetrix Rat Gene 1.0 ST Array 10775608 - -
          Affymetrix Rat Gene 1.0 ST Array 10775611 - -
          Affymetrix Rat Gene 1.0 ST Array 10775612 - -
          Affymetrix Rat Gene 1.0 ST Array 10775607 - -
          Affymetrix Rat Gene 1.0 ST Array 10775617 - -
          Affymetrix Rat Gene 1.0 ST Array 10775619 - -
          Affymetrix Rat Gene 1.0 ST Array 10775618 - -
          Affymetrix Rat Gene 1.0 ST Array 10775610 - -
          Affymetrix Rat Gene 1.0 ST Array 10775615 - -
          Affymetrix Rat Gene 1.0 ST Array 10775609 - -
          Affymetrix Rat Gene 1.0 ST Array 10775616 - -
          Affymetrix Rat Gene 1.0 ST Array 10775614 - -
          Affymetrix Rat Gene 1.1 ST Array 10775606 - -
          Rat Genome 230 2.0 Array 1379659_at - -
          Rat Genome 230 2.0 Array 1379659_PM_at - -
          Affymetrix HT Rat Focus Array 1379659_at - -
          Affymetrix HT Rat Focus Array 1379659_PM_at - -
          Rat Expression Set 230 1379659_PM_at - -
          Rat Expression Set 230 1379659_at - -
          Affymetrix Rat Gene 1.0 ST Array 10775606 - -
          Rat Genome U34 Set rc_AI012146_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366429 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372461 - -
          RatRef-12 Expression BeadChip ILMN_51657 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline