Search
Search
| Gene Symbol: Chdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198731.2 | NP_942026.1 | 4 - 5 | 1251 | 82 | |
| XM_006252578.2 | XP_006252640.1 | 4 - 5 | 1316 | 82 | ||
| XM_006252579.2 | XP_006252641.1 | 4 - 5 | 1270 | 82 | ||
| XM_006252580.2 | XP_006252642.1 | 4 - 5 | 1076 | 82 | ||
| XM_008770984.1 | XP_008769206.1 | 4 - 5 | 1619 | 82 | ||
| XM_008770985.1 | XP_008769207.1 | 5 - 6 | 1708 | 82 | ||
| GenBank mRNA | BC085787.1 | - | 4 - 5 | 1251 | 7108667 | 82 |
| X94769.1 | - | 3 - 4 | 526 | 82 | ||
Set Membership: |
|