Search
Search
| Gene Symbol: Pisd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001061408.5 | XP_001061408.2 | 6 - 7 | 868 | 103 | |
| XM_006251423.2 | XP_006251485.1 | 6 - 7 | 4798 | 103 | ||
| XM_006251424.2 | XP_006251486.1 | 6 - 7 | 3980 | 103 | ||
| XM_006251426.2 | XP_006251488.1 | 5 - 6 | 4562 | 103 | ||
| XM_008770389.1 | XP_008768611.1 | 5 - 6 | 4107 | 103 | ||
Set Membership: |
|