Search
Search
| Gene Symbol: Etv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228814.2 | XP_006228876.1 | 6 - 7 | 1700 | 84 | |
| XM_006228815.2 | XP_006228877.1 | 6 - 7 | 1718 | 84 | ||
| XM_008759255.1 | XP_008757477.1 | 5 - 6 | 1921 | 84 | ||
Set Membership: |
|