Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01460049_m1
          See other ZFP287 GE Assays ›
          Gene Symbol:
          Zfp287
          Gene Name:
          zinc finger protein 287
          Gene Aliases:
          Znf287
          Chromosome Location:
          Chr.10: 49132227 - 49150927 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01460049_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp287
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107008.2 NP_001100478.1 4 - 5 713 154
          XM_006246692.2 XP_006246754.1 5 - 6 1133 154

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002983.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of cytokine biosynthetic process
          positive regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P480939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448671 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44066 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79586 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480939 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480939 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P480939 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P480939 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10743605 - -
          Affymetrix Rat Gene 1.0 ST Array 10743601 - -
          Affymetrix Rat Gene 1.0 ST Array 10743604 - -
          Affymetrix Rat Gene 1.0 ST Array 10743602 - -
          Affymetrix Rat Gene 1.0 ST Array 10743603 - -
          Affymetrix Rat Gene 1.1 ST Array 10743600 - -
          Rat Genome 230 2.0 Array 1392574_at - -
          Rat Genome 230 2.0 Array 1392574_PM_at - -
          Rat Genome 230 2.0 Array 1385379_at - -
          Rat Genome 230 2.0 Array 1385379_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000026406.rn.1 TC1000002983.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000026409.rn.1 TC1000002983.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000026407.rn.1 TC1000002983.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000026408.rn.1 TC1000002983.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000026410.rn.1 TC1000002983.rn.1 RTA TC
          Rat Expression Set 230 1385379_at - -
          Rat Expression Set 230 1385379_PM_at - -
          Rat Expression Set 230 1392574_PM_at - -
          Rat Expression Set 230 1392574_at - -
          Affymetrix Rat Gene 1.0 ST Array 10743600 - -
          Rat Genome U34 Set rc_AI013120_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21951917 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357343 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline