Search
Search
| Gene Symbol: Mid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191889.1 | NP_001178818.1 | 4 - 5 | 1086 | 70 | |
| XM_006234247.2 | XP_006234309.1 | 4 - 5 | 1471 | 70 | ||
| XM_006234248.2 | XP_006234310.1 | 4 - 5 | 1027 | 70 | ||
Set Membership: |
|