Search
Search
| Gene Symbol: Ssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031119.2 | NP_112381.2 | 3 - 4 | 269 | 86 | |
| XM_006234321.2 | XP_006234383.1 | 3 - 4 | 345 | 86 | ||
| XM_006234322.2 | XP_006234384.1 | 3 - 4 | 345 | 86 | ||
| XM_006234323.2 | XP_006234385.1 | 3 - 4 | 345 | 86 | ||
| GenBank mRNA | BC081780.1 | - | 3 - 4 | 269 | 7192454 | 86 |
| BC097252.1 | - | 3 - 4 | 275 | 86 | ||
| X67859.1 | - | 2 - 3 | 204 | 86 | ||
Set Membership: |
|