Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01487500_m1
          See other RGD1564865 GE Assays ›
          Gene Symbol:
          RGD1564865
          Gene Name:
          similar to 20-alpha-hydroxysteroid dehydrogenase
          Gene Aliases:
          RGD1309566
          Chromosome Location:
          Chr.17: 69468427 - 69497132 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01487500_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1564865
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164396.1 NP_001157868.1 7 - 8 932 108

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          oxidation-reduction process
          oxidoreductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P102456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750994 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28345 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163880 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94689 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102456 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102456 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P102456 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1034879 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.1 ST Array 10796018 - -
          Affymetrix Rat Gene 1.1 ST Array 10860934 - -
          Affymetrix Rat Gene 1.1 ST Array 10796016 - -
          Rat Genome 230 2.0 Array 1393221_PM_at - -
          Rat Genome 230 2.0 Array 1393221_at - -
          Affymetrix HT Rat Focus Array 1393221_at - -
          Affymetrix HT Rat Focus Array 1393221_PM_at - -
          Rat Expression Set 230 1393221_PM_at - -
          Rat Expression Set 230 1393221_at - -
          Rat Genome U34 Set rc_AA891831_at - -
          Rat Genome U34 Set rc_AA866264_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22360783 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368626 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline