Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01493575_m1
          See other TMEM212 GE Assays ›
          Gene Symbol:
          Tmem212
          Gene Name:
          transmembrane protein 212
          Gene Aliases:
          RGD1563109
          Chromosome Location:
          Chr.2: 113578614 - 113616786 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01493575_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem212
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164438.1 NP_001157910.1 1 - 2 190 87
          NM_001164439.1 NP_001157911.1 1 - 2 190 87
          XM_006232219.2 XP_006232281.1 1 - 2 210 87
          XM_008760898.1 XP_008759120.1 1 - 2 210 87
          GenBank mRNA BE108013.1 - 3 - 4 87
          CO384351.1 - 1 - 2 190 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003390.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transporter

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P054692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109455 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131839 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92616 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122399 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6719 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054692 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054692 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P054692 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.1 ST Array 10822627 - -
          Rat Genome 230 2.0 Array 1378042_at - -
          Rat Genome 230 2.0 Array 1381901_at - -
          Rat Genome 230 2.0 Array 1381901_PM_at - -
          Rat Genome 230 2.0 Array 1378042_PM_at - -
          Affymetrix HT Rat Focus Array 1381901_PM_at - -
          Affymetrix HT Rat Focus Array 1381901_at - -
          Affymetrix HT Rat Focus Array 1378042_at - -
          Affymetrix HT Rat Focus Array 1378042_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200023196.rn.1 TC0200003390.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200023197.rn.1 TC0200003390.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200023198.rn.1 TC0200003390.rn.1 RTA TC
          Rat Expression Set 230 1381901_PM_at - -
          Rat Expression Set 230 1381901_at - -
          Rat Expression Set 230 1378042_PM_at - -
          Rat Expression Set 230 1378042_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21600583 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361549 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline