Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01500313_m1
          See other LOC100910708 GE Assays ›
          Gene Symbol:
          LOC100910708 Akr1b10
          Gene Name:
          aldose reductase-related protein 1-like
          aldo-keto reductase family 1, member B10 (aldose reductase)
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.4: 62015923 - 62031792 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 61813265 - 61828658 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01500313_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100910708
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008762861.1 XP_008761083.1 9 - 10 1233 114
          Gene Symbol: Akr1b10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013084.1 NP_001013102.1 10 - 11 1085 114
          GenBank mRNA BC079133.1 - 10 - 11 1085 7127413 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000619.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100910708
          Click here to find siRNAs for Gene Akr1b10

          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          oxidation-reduction process
          farnesol catabolic process
          polyol metabolic process
          retinol metabolic process
          retinal metabolic process
          daunorubicin metabolic process
          doxorubicin metabolic process
          retinal dehydrogenase activity
          alditol:NADP+ 1-oxidoreductase activity
          retinal binding
          geranylgeranyl reductase activity
          indanol dehydrogenase activity
          NADP-retinol dehydrogenase activity
          NADP+ binding
          NADPH binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P025608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322841 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14977 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132777 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59110 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025608 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P025608 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P025608 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P299977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100371 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65183 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77589 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10854422 - -
          Affymetrix Rat Gene 1.0 ST Array 10854420 - -
          Affymetrix Rat Gene 1.0 ST Array 10854424 - -
          Affymetrix Rat Gene 1.0 ST Array 10854425 - -
          Affymetrix Rat Gene 1.0 ST Array 10854419 - -
          Affymetrix Rat Gene 1.0 ST Array 10854421 - -
          Affymetrix Rat Gene 1.0 ST Array 10854426 - -
          Affymetrix Rat Gene 1.0 ST Array 10854418 - -
          Affymetrix Rat Gene 1.0 ST Array 10854423 - -
          Affymetrix Rat Gene 1.1 ST Array 10854417 - -
          Rat Genome 230 2.0 Array 1382034_at - -
          Rat Genome 230 2.0 Array 1395190_at - -
          Rat Genome 230 2.0 Array 1382034_PM_at - -
          Rat Genome 230 2.0 Array 1395190_PM_at - -
          Affymetrix HT Rat Focus Array 1395190_PM_at - -
          Affymetrix HT Rat Focus Array 1382034_PM_at - -
          Affymetrix HT Rat Focus Array 1382034_at - -
          Affymetrix HT Rat Focus Array 1395190_at - -
          Affymetrix Rat Clariom S Assay PSR0400004364.rn.1 TC0400000619.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400004363.rn.1 TC0400000619.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400004365.rn.1 TC0400000619.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400004366.rn.1 TC0400000619.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400004362.rn.1 TC0400000619.rn.1 RTA TC
          Rat Expression Set 230 1395190_PM_at - -
          Rat Expression Set 230 1382034_at - -
          Rat Expression Set 230 1382034_PM_at - -
          Rat Expression Set 230 1395190_at - -
          Affymetrix Rat Gene 1.0 ST Array 10854417 - -
          Rat Genome U34 Set rc_AI228017_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22041729 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351866 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline