Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01502867_m1
          See other ZDHHC17 GE Assays ›
          Gene Symbol:
          Zdhhc17
          Gene Name:
          zinc finger, DHHC-type containing 17
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 53653115 - 53714879 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01502867_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zdhhc17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039340.1 NP_001034429.1 14 - 15 1484 101
          GenBank mRNA AY886533.1 - 14 - 15 1484 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002690.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          magnesium ion transport
          protein palmitoylation
          lipoprotein transport
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          signal transducer activity
          zinc ion binding
          magnesium ion transmembrane transporter activity
          palmitoyltransferase activity
          protein-cysteine S-palmitoyltransferase activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P129346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841399 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50777 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77980 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146060 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91207 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7790 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170569 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63033 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129346 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129346 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P129346 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P282024 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21332 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85780 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67352 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77690 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7953 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17895 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10902301 - -
          Affymetrix Rat Gene 1.0 ST Array 10902311 - -
          Affymetrix Rat Gene 1.0 ST Array 10902295 - -
          Affymetrix Rat Gene 1.0 ST Array 10902307 - -
          Affymetrix Rat Gene 1.0 ST Array 10902304 - -
          Affymetrix Rat Gene 1.0 ST Array 10902306 - -
          Affymetrix Rat Gene 1.0 ST Array 10902298 - -
          Affymetrix Rat Gene 1.0 ST Array 10902312 - -
          Affymetrix Rat Gene 1.0 ST Array 10902302 - -
          Affymetrix Rat Gene 1.0 ST Array 10902299 - -
          Affymetrix Rat Gene 1.0 ST Array 10902300 - -
          Affymetrix Rat Gene 1.0 ST Array 10902310 - -
          Affymetrix Rat Gene 1.0 ST Array 10902297 - -
          Affymetrix Rat Gene 1.0 ST Array 10902308 - -
          Affymetrix Rat Gene 1.0 ST Array 10902309 - -
          Affymetrix Rat Gene 1.0 ST Array 10902303 - -
          Affymetrix Rat Gene 1.0 ST Array 10902296 - -
          Affymetrix Rat Gene 1.1 ST Array 10902294 - -
          Affymetrix Rat Clariom S Assay PSR0700020928.rn.1 TC0700002690.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700020930.rn.1 TC0700002690.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700020927.rn.1 TC0700002690.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700020929.rn.1 TC0700002690.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700020931.rn.1 TC0700002690.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10902294 - -
          Rat Genome U34 Set rc_AI072801_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline