Search
Search
| Gene Symbol: Atp2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053311.1 | NP_445763.1 | 13 - 14 | 2511 | 147 | |
| XM_006241294.2 | XP_006241356.1 | 13 - 14 | 2603 | 147 | ||
| XM_006241295.2 | XP_006241357.1 | 12 - 13 | 2480 | 147 | ||
| XM_006241296.2 | XP_006241358.1 | 13 - 14 | 2603 | 147 | ||
| GenBank mRNA | J03753.1 | - | 13 - 14 | 2511 | 147 | |
Set Membership: |
|