Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01510462_m1
          See other L7RN6 GE Assays ›
          Gene Symbol:
          l7Rn6
          Gene Name:
          lethal, Chr 7, Rinchik 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 154147098 - 154170428 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01510462_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: l7Rn6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013897.1 NP_001013919.1 4 - 5 660 90
          XM_006229662.2 XP_006229724.1 3 - 4 627 90
          XM_008759638.1 XP_008757860.1 3 - 4 458 90
          XM_008759639.1 XP_008757861.1 2 - 3 387 90
          GenBank mRNA BC088340.1 - 4 - 5 660 7367791 90
          FQ212130.1 - 3 - 4 571 90
          FQ213824.1 - 3 - 4 90
          FQ214677.1 - 3 - 4 572 90
          FQ215506.1 - 3 - 4 563 90
          FQ216197.1 - 3 - 4 572 90
          FQ216668.1 - 3 - 4 572 90
          FQ216759.1 - 3 - 4 572 90
          FQ218863.1 - 3 - 4 569 90
          FQ220044.1 - 3 - 4 572 90
          FQ220310.1 - 3 - 4 623 90
          FQ220575.1 - 3 - 4 90
          FQ224272.1 - 3 - 4 574 90
          FQ224945.1 - 3 - 4 572 90
          FQ229506.1 - 3 - 4 568 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006024.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene l7Rn6

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus
          Golgi organization
          protein transport
          lung development
          cellular response to heat
          protein transporter activity
          Hsp70 protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P096432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35686 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88819 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P096432 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P096432 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P096432 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P585359 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48974 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90981 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10723607 - -
          Affymetrix Rat Gene 1.0 ST Array 10723608 - -
          Affymetrix Rat Gene 1.0 ST Array 10723605 - -
          Affymetrix Rat Gene 1.0 ST Array 10723606 - -
          Affymetrix Rat Gene 1.0 ST Array 10723603 - -
          Affymetrix Rat Gene 1.0 ST Array 10723602 - -
          Affymetrix Rat Gene 1.0 ST Array 10723604 - -
          Affymetrix Rat Gene 1.1 ST Array 10723601 - -
          Rat Genome 230 2.0 Array 1371467_at - -
          Rat Genome 230 2.0 Array 1371467_PM_at - -
          Affymetrix HT Rat Focus Array 1371467_PM_at - -
          Affymetrix HT Rat Focus Array 1371467_at - -
          Affymetrix Rat Clariom S Assay PSR0100047882.rn.1 TC0100006024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100047877.rn.1 TC0100006024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100047878.rn.1 TC0100006024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100047880.rn.1 TC0100006024.rn.1 RTA TC
          Rat Expression Set 230 1371467_at - -
          Rat Expression Set 230 1371467_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10723601 - -
          Rat Genome U34 Set rc_AA891221_at - -
          Rat Genome U34 Set rc_AA998226_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21074400 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352305 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline