Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01512948_m1
          See other LOC691083 GE Assays ›
          Gene Symbol:
          LOC691083
          Gene Name:
          hypothetical protein LOC691083
          Gene Aliases:
          RGD1306990
          Chromosome Location:
          Chr.3: 95312809 - 95398594 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01512948_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC691083
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001144862.1 NP_001138334.1 277 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001322.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          esterase

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P004511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264102 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137808 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9249 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151372 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21131 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122451 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25310 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170241 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48082 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72600 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6101 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98707 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109302 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16992 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79986 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117946 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004511 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004511 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P004511 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P452600 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10838258 - -
          Affymetrix Rat Gene 1.0 ST Array 10838257 - -
          Affymetrix Rat Gene 1.1 ST Array 10838255 - -
          Rat Genome 230 2.0 Array 1391679_PM_at - -
          Rat Genome 230 2.0 Array 1391679_at - -
          Affymetrix HT Rat Focus Array 1391679_at - -
          Affymetrix HT Rat Focus Array 1391679_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300008988.rn.1 TC0300001322.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300008986.rn.1 TC0300001322.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300008987.rn.1 TC0300001322.rn.1 RTA TC
          Rat Expression Set 230 1391679_PM_at - -
          Rat Expression Set 230 1391679_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20804693 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline