Search
Search

Nek9| Gene Symbol: Nek9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106747.1 | NP_001100217.1 | 21 - 22 | 2948 | 62 | |
| XM_008764757.1 | XP_008762979.1 | 21 - 22 | 2972 | 62 | ||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P067055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145906 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P067055 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P067055 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P067055 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P127014 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P732126 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85713 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98659 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29363 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77330 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10891286 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891272 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891291 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891284 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891287 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891289 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891288 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891285 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891277 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891281 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891282 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891280 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891290 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891283 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891275 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891292 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891278 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10891271 | - | - |
| Rat Genome 230 2.0 Array | 1374040_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395341_at | - | - |
| Rat Genome 230 2.0 Array | 1390225_at | - | - |
| Rat Genome 230 2.0 Array | 1376454_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389617_at | - | - |
| Rat Genome 230 2.0 Array | 1374040_at | - | - |
| Rat Genome 230 2.0 Array | 1390225_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389617_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375271_at | - | - |
| Rat Genome 230 2.0 Array | 1375271_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376454_at | - | - |
| Rat Genome 230 2.0 Array | 1395341_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375271_at | - | - |
| Affymetrix HT Rat Focus Array | 1375271_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376454_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395341_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374040_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376454_at | - | - |
| Affymetrix HT Rat Focus Array | 1395341_at | - | - |
| Affymetrix HT Rat Focus Array | 1390225_at | - | - |
| Affymetrix HT Rat Focus Array | 1390225_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374040_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600020385.rn.1 | TC0600002869.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600020384.rn.1 | TC0600002869.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600020386.rn.1 | TC0600002869.rn.1 | RTA TC |
| Rat Expression Set 230 | 1375271_PM_at | - | - |
| Rat Expression Set 230 | 1376454_PM_at | - | - |
| Rat Expression Set 230 | 1375271_at | - | - |
| Rat Expression Set 230 | 1390225_PM_at | - | - |
| Rat Expression Set 230 | 1376454_at | - | - |
| Rat Expression Set 230 | 1374040_PM_at | - | - |
| Rat Expression Set 230 | 1390225_at | - | - |
| Rat Expression Set 230 | 1374040_at | - | - |
| Rat Expression Set 230 | 1395341_PM_at | - | - |
| Rat Expression Set 230 | 1395341_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10891271 | - | - |
| Rat Genome U34 Set | rc_AI045249_at | - | - |
| Rat Genome U34 Set | rc_AA901069_at | - | - |
| Rat Genome U34 Set | rc_AA901069_g_at | - | - |
| Rat Genome U34 Set | rc_AA818725_at | - | - |
| Rat Genome U34 Set | rc_AI102451_at | - | - |
| Rat Genome U34 Set | rc_AI071673_at | - | - |
| Rat Genome U34 Set | rc_AI045249_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22309999 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371075 | - | - |