Search
Search
| Gene Symbol: Sgol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766787.1 | XP_008765009.1 | 7 - 8 | 1534 | 92 | |
| XM_008766788.1 | XP_008765010.1 | 7 - 8 | 1503 | 92 | ||
| XM_008766789.1 | XP_008765011.1 | 7 - 8 | 1531 | 92 | ||
Set Membership: |
|