Search
Search
| Gene Symbol: Vamp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249016.2 | XP_006249078.1 | 7 - 8 | 743 | 143 | |
| GenBank mRNA | FQ228923.1 | - | 7 - 8 | 703 | 143 | |
| FQ230649.1 | - | 6 - 7 | 514 | 143 | ||
Set Membership: |
|