Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01531696_m1
          See other RSPRY1 GE Assays ›
          Gene Symbol:
          Rspry1
          Gene Name:
          ring finger and SPRY domain containing 1
          Gene Aliases:
          RGD1308847
          Chromosome Location:
          Chr.19: 10770572 - 10826895 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01531696_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rspry1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100945.1 NP_001094415.1 14 - 15 2091 58
          GenBank mRNA AY724539.1 - 13 - 14 1734 58
          BC129112.1 - 14 - 15 2091 7112017 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000998.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157638 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P155334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124923 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150623 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110899 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157638 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155334 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155334 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157638 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P155334 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157638 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1021456 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64171 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23671 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36289 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20039 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10809291 - -
          Affymetrix Rat Gene 1.0 ST Array 10809287 - -
          Affymetrix Rat Gene 1.0 ST Array 10809294 - -
          Affymetrix Rat Gene 1.0 ST Array 10809293 - -
          Affymetrix Rat Gene 1.0 ST Array 10809301 - -
          Affymetrix Rat Gene 1.0 ST Array 10809290 - -
          Affymetrix Rat Gene 1.0 ST Array 10809288 - -
          Affymetrix Rat Gene 1.0 ST Array 10809292 - -
          Affymetrix Rat Gene 1.0 ST Array 10809300 - -
          Affymetrix Rat Gene 1.0 ST Array 10809289 - -
          Affymetrix Rat Gene 1.0 ST Array 10809298 - -
          Affymetrix Rat Gene 1.0 ST Array 10809299 - -
          Affymetrix Rat Gene 1.0 ST Array 10809297 - -
          Affymetrix Rat Gene 1.0 ST Array 10809295 - -
          Affymetrix Rat Gene 1.0 ST Array 10809296 - -
          Affymetrix Rat Gene 1.1 ST Array 10809286 - -
          Rat Genome 230 2.0 Array 1379221_PM_at - -
          Rat Genome 230 2.0 Array 1379221_at - -
          Rat Genome 230 2.0 Array 1385860_at - -
          Rat Genome 230 2.0 Array 1385860_PM_at - -
          Affymetrix HT Rat Focus Array 1385860_at - -
          Affymetrix HT Rat Focus Array 1385860_PM_at - -
          Affymetrix HT Rat Focus Array 1379221_at - -
          Affymetrix HT Rat Focus Array 1379221_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900007963.rn.1 TC1900000998.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007962.rn.1 TC1900000998.rn.1 RTA TC
          Rat Expression Set 230 1385860_PM_at - -
          Rat Expression Set 230 1385860_at - -
          Rat Expression Set 230 1379221_at - -
          Rat Expression Set 230 1379221_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10809286 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline