Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01757120_m1
          See other CYP39A1 GE Assays ›
          Gene Symbol:
          Cyp39a1
          Gene Name:
          cytochrome P450, family 39, subfamily a, polypeptide 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 19804079 - 19880346 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01757120_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp39a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106893.1 NP_001100363.1 3 - 4 807 70
          XM_006244604.2 XP_006244666.1 3 - 4 807 70
          XM_006244605.1 XP_006244667.1 3 - 4 807 70
          XM_008766854.1 XP_008765076.1 3 - 4 690 70
          XM_008766855.1 XP_008765077.1 3 - 4 688 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001653.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          bile acid biosynthetic process
          cholesterol catabolic process
          biological_process
          oxidation-reduction process
          molecular_function
          iron ion binding
          steroid 7-alpha-hydroxylase activity
          oxysterol 7-alpha-hydroxylase activity
          heme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P229837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952905 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54015 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127517 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147234 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90457 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72339 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53830 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153417 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114890 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P229837 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P229837 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P229837 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P229837 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36057 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39409 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10926676 - -
          Affymetrix Rat Gene 1.0 ST Array 10926670 - -
          Affymetrix Rat Gene 1.0 ST Array 10926672 - -
          Affymetrix Rat Gene 1.0 ST Array 10926674 - -
          Affymetrix Rat Gene 1.0 ST Array 10926679 - -
          Affymetrix Rat Gene 1.0 ST Array 10926675 - -
          Affymetrix Rat Gene 1.0 ST Array 10926669 - -
          Affymetrix Rat Gene 1.0 ST Array 10926678 - -
          Affymetrix Rat Gene 1.0 ST Array 10926680 - -
          Affymetrix Rat Gene 1.0 ST Array 10926671 - -
          Affymetrix Rat Gene 1.0 ST Array 10926677 - -
          Affymetrix Rat Gene 1.0 ST Array 10926673 - -
          Affymetrix Rat Gene 1.1 ST Array 10926668 - -
          Rat Genome 230 2.0 Array 1385165_at - -
          Rat Genome 230 2.0 Array 1385165_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900013589.rn.1 TC0900001653.rn.1 RTA TC
          Rat Expression Set 230 1385165_at - -
          Rat Expression Set 230 1385165_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10926668 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21401352 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353886 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline