Search
Search
| Gene Symbol: Fbxo28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107203.3 | NP_001100673.1 | 4 | 621 | 136 | |
| GenBank mRNA | BC167003.1 | - | 4 | 622 | 8366732 | 136 |
| FQ223120.1 | - | 3 | 317 | 136 | ||
Set Membership: |
|