Search
Search
| Gene Symbol: Glyat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009648.2 | NP_001009648.1 | 1 - 2 | 74 | 137 | |
| XM_006231121.2 | XP_006231183.1 | 3 - 4 | 381 | 137 | ||
| XM_006231122.2 | XP_006231184.1 | 2 - 3 | 296 | 137 | ||
| XM_006231123.2 | XP_006231185.1 | 2 - 3 | 118 | 137 | ||
| XM_008760257.1 | XP_008758479.1 | 3 - 4 | 1261 | 137 | ||
| GenBank mRNA | AB201944.1 | - | 1 - 2 | 74 | 137 | |
| AY325192.1 | - | 2 - 3 | 114 | 137 | ||
| BC087043.1 | - | 2 - 3 | 172 | 7108337 | 137 | |
Set Membership: |
|