Search
Search

Ppia
LOC100360977| Gene Symbol: Ppia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017101.1 | NP_058797.1 | 3 - 4 | 233 | 70 | |
| GenBank mRNA | BC059141.1 | - | 3 - 4 | 238 | 6919594 | 70 |
| BC091153.1 | - | 3 - 4 | 209 | 7308693 | 70 | |
| FQ210268.1 | - | 3 - 4 | 234 | 70 | ||
| FQ210843.1 | - | 3 - 4 | 236 | 70 | ||
| FQ222826.1 | - | 3 - 4 | 234 | 70 | ||
| FQ223366.1 | - | 3 - 4 | 234 | 70 | ||
| FQ223500.1 | - | 3 - 4 | 234 | 70 | ||
| FQ227889.1 | - | 3 - 4 | 234 | 70 | ||
| FQ228411.1 | - | 3 - 4 | 234 | 70 | ||
| FQ228450.1 | - | 3 - 4 | 234 | 70 | ||
| FQ231674.1 | - | 3 - 4 | 234 | 70 | ||
| FQ233894.1 | - | 3 - 4 | 233 | 70 | ||
| FQ234872.1 | - | 3 - 4 | 234 | 70 | ||
| M19533.1 | - | 3 - 4 | 233 | 70 | ||
| M25637.1 | - | 3 - 4 | 226 | 70 | ||
| Gene Symbol: LOC100360977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250801.2 | XP_006250863.1 | 1 | 230 | 70 | |
Set Membership: |
|