Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04223554_gH
          See other CLEC2D2 GE Assays ›
          Gene Symbol:
          Clec2d2 Clec2dl1 LOC689757
          Gene Name:
          C-type lectin domain family 2 member D2
          C-type lectin domain family 2 member D-like 1
          similar to osteoclast inhibitory lectin
          Gene Aliases:
          Clr2, RGD1562831
          Clec2d6
          Clec2d3, RGD1563033
          Chromosome Location:
          Chr.4: 162437099 - 162445285 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 162391376 - 162402569 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 162181798 - 162229409 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn04223554_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clec2d2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085402.1 NP_001078871.1 3 - 4 462 94
          GenBank mRNA EF100686.1 - 3 - 4 462 94
          EU128746.1 - 3 - 4 468 94
          FQ221935.1 - 553 94
          Gene Symbol: Clec2dl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085404.1 NP_001078873.1 3 - 4 462 94
          XM_006237453.2 XP_006237515.1 4 - 5 495 94
          XM_008763333.1 XP_008761555.1 3 - 4 1521 94
          GenBank mRNA EF100689.1 - 3 - 4 462 94
          FJ404471.1 - 3 - 4 462 94
          Gene Symbol: LOC689757
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001101019.2 NP_001094489.2 3 - 4 546 94
          XM_008763332.1 XP_008761554.1 3 - 4 849 94
          GenBank mRNA FJ404472.1 - 3 - 4 546 94

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Process(es)

          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P069365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588886 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069365 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069365 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P069365 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10865959 - -
          Affymetrix Rat Gene 1.0 ST Array 10865958 - -
          Affymetrix Rat Gene 1.0 ST Array 10865957 - -
          Affymetrix Rat Gene 1.1 ST Array 10865956 - -
          Affymetrix Rat Clariom S Assay PSR0400013576.rn.1 TC0400001865.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027315.rn.1 TC0400003836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400027316.rn.1 TC0400003836.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400013573.rn.1 TC0400001865.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400013587.rn.1 TC0400001865.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400013574.rn.1 TC0400001865.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400013586.rn.1 TC0400001865.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400013572.rn.1 TC0400001865.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400013575.rn.1 TC0400001865.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400013588.rn.1 TC0400001865.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10865956 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21017602 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361801 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline