Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn99999011_m1
See other IL6 GE Assays ›
Gene Symbol:
Il6
Gene Name:
interleukin 6
Gene Aliases:
ILg6, Ifnb2
Chromosome Location:
Chr.4: 3043231 - 3047807 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn99999011_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Il6
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_012589.2
NP_036721.1
2 - 3
268
90
GenBank mRNA
M26744.1
-
2 - 3
268
90
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0400000061.rn.1
Set Membership:
Ambion siRNA available
Amplicon length between 86 and 100
Apoptosis
Rattus norvegicus
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Il6
Gene Ontology Categories:
Function(s)
Process(es)
neutrophil apoptotic process
response to yeast
myeloid cell homeostasis
hepatic immune response
positive regulation of acute inflammatory response
negative regulation of protein kinase activity
acute-phase response
inflammatory response
aging
positive regulation of cell proliferation
negative regulation of cell proliferation
regulation of cell shape
response to heat
response to cold
response to wounding
response to mechanical stimulus
response to bacterium
regulation of vascular endothelial growth factor production
positive regulation of gene expression
response to auditory stimulus
response to organic cyclic compound
cell growth
cytokine-mediated signaling pathway
response to caffeine
endocrine pancreas development
neuron projection development
response to nutrient levels
response to peptidoglycan
response to lipopolysaccharide
positive regulation of chemokine production
positive regulation of interleukin-6 production
response to insulin
negative regulation of collagen biosynthetic process
positive regulation of peptidyl-serine phosphorylation
positive regulation of protein import into nucleus, translocation
response to cytokine
wound healing
positive regulation of T cell proliferation
T cell activation
regulation of cell proliferation
response to drug
positive regulation of tyrosine phosphorylation of Stat3 protein
glucose homeostasis
defense response to protozoan
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to amino acid
positive regulation of MAPK cascade
negative regulation of chemokine biosynthetic process
regulation of circadian sleep/wake cycle, non-REM sleep
positive regulation of nitric oxide biosynthetic process
cell redox homeostasis
positive regulation of T-helper 2 cell differentiation
positive regulation of neuron differentiation
negative regulation of gluconeogenesis
positive regulation of translation
positive regulation of DNA replication
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JAK-STAT cascade
response to antibiotic
muscle cell cellular homeostasis
bone remodeling
negative regulation of hormone secretion
negative regulation of muscle organ development
positive regulation of smooth muscle cell proliferation
positive regulation of epithelial cell proliferation
negative regulation of cytokine secretion
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
positive regulation of B cell activation
positive regulation of immunoglobulin secretion
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
response to glucocorticoid
response to calcium ion
response to electrical stimulus
defense response to virus
positive regulation of protein kinase B signaling
positive regulation of transmission of nerve impulse
branching involved in salivary gland morphogenesis
epithelial cell proliferation involved in salivary gland morphogenesis
glucagon secretion
interleukin-6-mediated signaling pathway
cellular response to hydrogen peroxide
positive regulation of ERK1 and ERK2 cascade
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to dexamethasone stimulus
T-helper 17 cell lineage commitment
negative regulation of neuron death
receptor binding
cytokine activity
interleukin-6 receptor binding
growth factor activity
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR0400000330.rn.1
TC0400000061.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline