Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Sc04111346_s1
See other SAC3 GE Assays ›
Gene Symbol:
SAC3
Gene Name:
Gene Aliases:
LEP1, YDR159W
Chromosome Location:
Chr.IV: 771877 - 775782 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID
Sc04111346_s1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: SAC3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001180466.3
NP_010443.3
1
3652
78
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Detect Genomic DNA
Saccharomyces cerevisiae
Both primers and probe map within a single exon
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cell cycle
posttranscriptional tethering of RNA polymerase II gene DNA at nuclear periphery
transcription-coupled nucleotide-excision repair
mRNA export from nucleus
protein export from nucleus
actin filament-based process
mRNA 3'-end processing
ribosomal small subunit biogenesis
mRNA transport
nuclear mRNA surveillance
protein binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline