Search
Search
| Gene Symbol: FDX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214065.1 | NP_999230.1 | 4 | 453 | 74 | |
| GenBank mRNA | AK400648.1 | - | 4 | 612 | 74 | |
| AY610208.1 | - | 4 | 661 | 74 | ||
| M57674.1 | - | 4 | 453 | 74 | ||
Set Membership: |
|