Search
Search
| Gene Symbol: PPARG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214379.1 | NP_999544.1 | 6 - 7 | 1285 | 72 | |
| XM_005669783.3 | XP_005669840.1 | 1381 | 72 | |||
| XM_005669784.3 | XP_005669841.1 | 1442 | 72 | |||
| XM_005669788.3 | XP_005669845.3 | 1298 | 72 | |||
| XM_005669789.3 | XP_005669846.1 | 1369 | 72 | |||
| XM_013981980.2 | XP_013837434.1 | 1215 | 72 | |||
| XM_013981981.2 | XP_013837435.1 | 1660 | 72 | |||
| XM_013981982.2 | XP_013837436.1 | 1558 | 72 | |||
| GenBank mRNA | AB097926.1 | - | 7 - 8 | 1345 | 72 | |
| AB097927.1 | - | 6 - 7 | 1271 | 72 | ||
| AB097928.1 | - | 7 - 8 | 1338 | 72 | ||
| AB097929.1 | - | 6 - 7 | 1264 | 72 | ||
| AB097930.1 | - | 6 - 7 | 1306 | 72 | ||
| AF059245.1 | - | 6 - 7 | 1285 | 72 | ||
| AF103946.1 | - | 6 - 7 | 1284 | 72 | ||
| AK230961.1 | - | 6 - 7 | 1390 | 72 | ||
| AK389674.1 | - | 6 - 7 | 1413 | 72 | ||
| DQ437884.1 | - | 7 - 8 | 1337 | 72 | ||
| DQ437885.1 | - | 6 - 7 | 1307 | 72 | ||
| KM382175.1 | - | 5 - 6 | 1182 | 72 | ||
Set Membership: |
|