Search
Search
| Gene Symbol: FSCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146300.2 | NP_001139772.1 | 4 - 5 | 1409 | 54 | |
| GenBank mRNA | AK236071.1 | - | 1 - 2 | 147 | 54 | |
| AK236699.1 | - | 4 - 5 | 1411 | 54 | ||
| AK392031.1 | - | 4 - 5 | 1407 | 54 | ||
| FJ790418.1 | - | 4 - 5 | 1411 | 54 | ||
Set Membership: |
|