Search
Search

| Gene Symbol: SCARB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244155.1 | NP_001231084.1 | 4 - 5 | 929 | 75 | |
| XM_005666776.2 | XP_005666833.1 | 924 | 75 | |||
| GenBank mRNA | AK236346.1 | - | 4 - 5 | 916 | 75 | |
| AK395497.1 | - | 4 - 5 | 929 | 75 | ||
| AY609583.1 | - | 4 - 5 | 728 | 75 | ||
| Gene Symbol: AASDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK391323.1 | - | 2669 | 75 | ||
Set Membership: |
|