Search
Search

srsf3| Gene Symbol: srsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005054.1 | NP_001005054.1 | 3 - 4 | 416 | 79 | |
| XM_012960059.2 | XP_012815513.1 | 3 - 4 | 712 | 79 | ||
| XM_012960068.2 | XP_012815522.1 | 3 - 4 | 716 | 79 | ||
| XM_012960072.2 | XP_012815526.1 | 3 - 4 | 481 | 79 | ||
| GenBank mRNA | BC076942.1 | - | 3 - 4 | 416 | 79 | |
| CR761747.2 | - | 3 - 4 | 374 | 79 | ||
| CT025261.2 | - | 3 - 4 | 371 | 79 | ||
Set Membership: |
|